missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR97 Antibody (2435C) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10097JF669
This item is not returnable.
View return policy
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| GPR97 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
| Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession # Q86Y34 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 2435C | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| GPCR | |
| 222487 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction