missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR15L Antibody (1020904), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB104161S
This item is not returnable.
View return policy
Description
GPR15L Monoclonal antibody specifically detects GPR15L in Human samples. It is validated for ELISA, Flow Cytometry, CyTOF
Specifications
| GPR15L | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 387695 | |
| Store at 4C in the dark. | |
| IgG2b |
| ELISA, Flow Cytometry, CyTOF | |
| 1020904 | |
| ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| C10orf99 chromosome 10 open reading frame 99, UNQ1833 | |
| E. coli-derived human C10orf99 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction