missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Golgi Glycoprotein 1/GLG1 Antibody (GLG1/970), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47794JF549
This item is not returnable.
View return policy
Description
Golgi Glycoprotein 1/GLG1 Monoclonal antibody specifically detects Golgi Glycoprotein 1/GLG1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Golgi Glycoprotein 1/GLG1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| CFR1, CFR-1, cysteine-rich fibroblast growth factor receptor, DKFZp686L08213, E-selectin ligand 1, ESL1, ESL-1, FLJ23319, FLJ23967, FLJ32797, Golgi apparatus protein 1, golgi glycoprotein 1, golgi sialoglycoprotein MG-160, MG160, MG-160 | |
| Golgi fraction from human liver cells | |
| 0.1 mL | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GLG1/970 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2734 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction