missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GGT1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98438CL1
This item is not returnable.
View return policy
Description
GGT1 Polyclonal antibody specifically detects GGT1 in Rat samples. It is validated for ELISA,Immunohistochemistry (Paraffin)
Specifications
| GGT1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| CD224, CD224 antigen, D22S672, D22S732, EC 2.3.2.2, gamma-glutamyl transpeptidase, gamma-glutamyltransferase 1MGC96904, gamma-glutamyltranspeptidase 1, GGT 1, GGTMGC96963, glutamyl transpeptidase, GTG, MGC96892 | |
| Produced in rabbits immunized with purified, recombinant Rat GGT1 (Uniprot#: P07314; Pro27-Tyr568). | |
| 0.1 mL | |
| Cancer | |
| 2678 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction