missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-B R2 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-159MFV450
This item is not returnable.
View return policy
Description
GABA-B R2 Polyclonal antibody specifically detects GABA-B R2 in Human samples. It is validated for Western Blot
Specifications
| GABA-B R2 | |
| Polyclonal | |
| Western Blot | |
| G protein-coupled receptor 51, GABA-BR2, GABA-B-R2, GABABR2GABA-B receptor 2, gamma-aminobutyric acid (GABA) B receptor, 2, gamma-aminobutyric acid type B receptor subunit 2, Gb2, GPR51GABA-B receptor, R2 subunit, GPRC3BFLJ36928, G-protein coupled receptor 51, HG20HRIHFB2099 | |
| Synthetic peptide made to the 105 kDa GABAbR2 protein. | |
| 0.1 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific | |
| 9568 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction