missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-7 Antibody (151143), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1981L
This item is not returnable.
View return policy
Description
Frizzled-7 Monoclonal antibody specifically detects Frizzled-7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Frizzled-7 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 151143 | |
| Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Fz-7, FzE3frizzled-7, hFz7 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7, Gln33-Leu185, Accession # Q61090 | |
| 0.1 mL | |
| GPCR, Growth and Development, Neuronal Cell Markers, Signal Transduction, Wnt Signaling Pathway | |
| 8324 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction