missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB2 Antibody (PCRP-FOXB2-2B2) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14125JF525
This item is not returnable.
View return policy
Description
FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array
Specifications
| FOXB2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| bA159H20.4, forkhead box B2, forkhead box protein B2 | |
| Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array | |
| PCRP-FOXB2-2B2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 442425 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction