missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC2 Antibody (04), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06212CL1
This item is not returnable.
View return policy
Description
DYDC2 Monoclonal antibody specifically detects DYDC2 in Human samples. It is validated for ELISA
Specifications
| DYDC2 | |
| Monoclonal | |
| CoraFluor 1 | |
| bA36D19.6, DPY30 domain containing 2, DPY30 domain-containing protein 2, MGC16186 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DYDC2 (Uniprot#: Q96IM9; Thr102-Pro176). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 04 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 84332 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction