missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DMRT2 Antibody (PCRP-DMRT2-1B11) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14270JF585
This item is not returnable.
View return policy
Description
DMRT2 Monoclonal antibody specifically detects DMRT2 in Human samples. It is validated for Flow Cytometry,Western Blot,Protein Array
Specifications
| DMRT2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| doublesex and mab-3 related transcription factor 2, doublesex- and mab-3-related transcription factor 2, Doublesex-like 2 protein, DSXL2, DSXL-2, terra-like protein | |
| Recombinant full-length human DMRT2 protein (Uniprot: Q9Y5R5) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Western Blot, Protein Array | |
| PCRP-DMRT2-1B11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 10655 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction