missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI12D7), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72227APC
This item is not returnable.
View return policy
Description
dedicator of cytokinesis 8 Monoclonal antibody specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| dedicator of cytokinesis 8 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI12D7 | |
| Western Blot, Immunohistochemistry | |
| dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
| Human recombinant protein fragment corresponding to amino acids 84-405 of human DOCK8 (NP_982272) produced in E.coli. | |
| 0.1 mL | |
| Cardiovascular Biology, Signal Transduction | |
| 81704 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction