missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDT Antibody (OTI1H3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70562APC
This item is not returnable.
View return policy
Description
DDT Monoclonal antibody specifically detects DDT in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DDT | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1H3 | |
| Western Blot, Immunohistochemistry | |
| D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II | |
| Full length human recombinant protein of human DDT(NP_001346) produced in HEK293T cell. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1652 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction