missing translation for 'onlineSavingsMsg'
Få mere at vide

Cytokeratin 3/2p Antibody (2Q1040(AE5)), Alexa Fluor™ 405, Novus Biologicals™

Artikelnummer. 18730173 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
0.1 mL
Pakningsstørrelse:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
18730173 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18730173 Leverandør Novus Biologicals Leverandørnr. NBP234844AF405

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

Mouse Monoclonal Antibody

Cytokeratin 3/2p Monoclonal antibody specifically detects Cytokeratin 3/2p in Human, Mouse, Bovine, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen Cytokeratin 3/2p
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2Q1040(AE5)
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias 65 KDa Cytokeratin, CK3, CK-3, cytokeratin-3, Keratin, keratin 3, Keratin, Type II Cytoskeletal 3, Keratin-3, Type II Cytoskeletal 3, Type-II keratin Kb3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratin
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3850
Target Species Human, Mouse, Bovine, Rabbit
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.