missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin B2 Antibody (X29.2), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08843CL1
This item is not returnable.
View return policy
Description
Cyclin B2 Monoclonal antibody specifically detects Cyclin B2 in Human,Mouse,Rat,Xenopus samples. It is validated for ELISA
Specifications
| Cyclin B2 | |
| Monoclonal | |
| CoraFluor 1 | |
| cyclin B2, G2/mitotic-specific cyclin-B2, HsT17299 | |
| Recombinant full-length Xenopus laevis Cyclin B2 protein (Uniprot: O95067) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 9133 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| ELISA | |
| X29.2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Xenopus species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction