missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL5/ENA-78 Antibody (MM0216-10L19), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12231MFV500
This item is not returnable.
View return policy
Description
CXCL5/ENA-78 Monoclonal antibody specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Specifications
| CXCL5/ENA-78 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization | |
| MM0216-10L19 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize | |
| chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
| Human recombinant CXCL5/ENA-78 | |
| 0.1 mL | |
| Biologically Active Proteins, Cell Cycle and Replication, Chemokines and Cytokines | |
| 6374 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction