missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CORO7 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18450JF525
This item is not returnable.
View return policy
Description
CORO7 Polyclonal antibody specifically detects CORO7 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CORO7 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79585 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 525 | |
| coronin-7, 0610011B16Rik, coronin 7, CRN7, POD1 | |
| Recognizes region between residues Between Between 400 and 450.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion