missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Coagulation Factor VII Antibody (321605), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB23381PEATT594
This item is not returnable.
View return policy
Description
Coagulation Factor VII Monoclonal antibody specifically detects Coagulation Factor VII in Human samples. It is validated for Flow Cytometry
Specifications
| Coagulation Factor VII | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 321605 | |
| Flow Cytometry | |
| coagulation factor VII, coagulation factor VII (serum prothrombin conversion accelerator), EC 3.4.21, EC 3.4.21.21, Eptacog alfa, FVII coagulation protein, Proconvertin, Serum prothrombin conversion accelerator, SPCA | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor VII, Ala39-Pro444, Accession # NP_062562 | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 2155 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction