missing translation for 'onlineSavingsMsg'
Learn More

Claudin-6 Antibody (AB3-7) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30160632
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30160632 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30160632 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328480JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

Claudin-6 Monoclonal antibody specifically detects Claudin-6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin-6
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone AB3-7
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias claudin 6, claudin-6, skullin
Host Species Human
Immunogen CLDN6
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9074
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.