missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Claudin-18.2 Antibody (CLDN18.2/7300R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24134JF635
This item is not returnable.
View return policy
Description
Claudin-18.2 Monoclonal antibody specifically detects Claudin-18.2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Claudin-18.2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| claudin-18, SFTA5, SFTPJ, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J | |
| Recombinant fragment (around aa1-100) of human Claudin-18.2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 51208 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| CLDN18.2/7300R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction