missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CKAP1 Antibody (1F6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-31207CL1
This item is not returnable.
View return policy
Description
CKAP1 Monoclonal antibody specifically detects CKAP1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| CKAP1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CG22Tubulin-specific chaperone B, CKAP1MGC14625, CKAPI, cytoskeleton associated protein 1, Cytoskeleton-associated protein 1Cytoskeleton-associated protein CKAPI, tubulin folding cofactor B, tubulin-folding cofactor B | |
| Recombinant human CKAP1 (1-244aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1F6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1155 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction