missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCP1 Antibody (U.California patent anti-CDCP1) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28720JF635
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal antibody specifically detects CDCP1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CDCP1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| CDCP1/ CD318 | |
| 0.1 mL | |
| Phospho Specific | |
| 64866 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| U.California patent anti-CDCP1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction