missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCP1 Antibody (469114), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB45151MFV610
This item is not returnable.
View return policy
Description
CDCP1 Monoclonal antibody specifically detects CDCP1 in Mouse samples. It is validated for Western Blot
Specifications
| CDCP1 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 469114 | |
| Western Blot | |
| CD318, CD318 antigen, CUB domain containing protein 1, CUB domain-containing protein 1, Membrane glycoprotein gp140, SIMA135TRASK, Subtractive immunization M plus HEp3-associated 135 kDa protein, Transmembrane and associated with src kinases | |
| Mouse myeloma cell line NS0-derived recombinant mouse CDCP1, Arg25-Ala667, Accession # Q5U462 | |
| 0.1 mL | |
| Phospho Specific | |
| 64866 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction