missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD7 Antibody (Persongen patent anti-CD7) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28213JF669
This item is not returnable.
View return policy
Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| CD7 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| CD7 antigen, CD7 antigen (p41), CD7 molecule, GP40T-cell surface antigen Leu-9, LEU-9, T-cell antigen CD7, T-cell leukemia antigen, Tp40, TP41p41 protein | |
| CD7 | |
| 0.1 mL | |
| Cytokine Research, Signal Transduction | |
| 924 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Persongen patent anti-CD7 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction