missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (CR1/802), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47865AF700
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human, Monkey, Baboon samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| CD35 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Recombinant full-length human CD35 protein (Uniprot: P17927) | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| CR1/802 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Monkey, Baboon | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction