missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD3 delta Polyclonal antibody specifically detects CD3 delta in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
Specifications
Specifications
| Classification | Polyclonal |
| Conjugate | APC |
| Gene ID (Entrez) | 915 |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?