missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1e Antibody (704407) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7330JF525
This item is not returnable.
View return policy
Description
CD1e Monoclonal antibody specifically detects CD1e in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| CD1e | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD1e molecule, e polypeptide, FLJ17609, membrane-associated, thymocyte antigen CD1E | |
| Mouse myeloma cell line NS0-derived recombinant human CD1e, Met1-Tyr304, Accession # P15812 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 704407 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Immunology | |
| 913 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction