missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 2 Antibody (OTI3G1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70334DL594
This item is not returnable.
View return policy
Description
Calpain 2 Monoclonal antibody specifically detects Calpain 2 in Human, Mouse, Rat, Canine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Calpain 2 | |
| Monoclonal | |
| DyLight 594 | |
| Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain | |
| Full length recombinant protein of human Calpain 2 (NP_001739) produced in HEK293T cells. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Cell Biology, Endocrinology, Immunology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 824 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI3G1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction