missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNG4 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99568FR
This item is not returnable.
View return policy
Description
CACNG4 Polyclonal antibody specifically detects CACNG4 in Human samples. It is validated for Western Blot
Specifications
| CACNG4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 680 | |
| calcium channel, voltage-dependent, gamma subunit 4, MGC11138, MGC24983, neuronal voltage-gated calcium channel gamma-4 subunit, TARP gamma-4, transmembrane AMPAR regulatory protein gamma-4, voltage-dependent calcium channel gamma-4 subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CACNG4. | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 27092 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction