missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRCA1 Antibody (MU), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-600JF549
This item is not returnable.
View return policy
Description
BRCA1 Monoclonal antibody specifically detects BRCA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation
Specifications
| BRCA1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| BRCAI, breast and ovarian cancer susceptibility protein 1, breast and ovarian cancer sususceptibility protein, breast cancer 1, early onset, breast cancer type 1 susceptibility protein, EC 6.3.2, EC 6.3.2.-, IRIS, PNCA4, PSCP, subunit 1 | |
| Human BRCA1 corresponding to residues 1314-1864 [UniProt# P38398] | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Cell Biology, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
| 672 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunoprecipitation | |
| MU | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction