missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLMH/Bleomycin Hydrolase Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98334CL1
This item is not returnable.
View return policy
Description
BLMH/Bleomycin Hydrolase Polyclonal antibody specifically detects BLMH/Bleomycin Hydrolase in Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| BLMH/Bleomycin Hydrolase | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| BHBMHBLM hydrolase, bleomycin hydrolase, EC 3.4.22.40 | |
| Produced in rabbits immunized with purified, recombinant Mouse BLMH/Bleomycin Hydrolase (Accession#: NP_848760.1; Asn 2-Glu 455) | |
| 0.1 mL | |
| Cancer, DNA Repair, Genes Sensitive to DNA Damaging Agents | |
| 642 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction