missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclin 2 Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB110-60982JF525
This item is not returnable.
View return policy
Description
Beclin 2 Polyclonal antibody specifically detects Beclin 2 in Human samples. It is validated for Western Blot
Specifications
| Beclin 2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 525 | |
| BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 | |
| A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot # A8MW95] | |
| 0.1 mL | |
| Apoptosis, Autophagy, Cancer, Tumor Suppressors | |
| 441925 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction