missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBS4 Antibody (OTI2D5), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70241APC
This item is not returnable.
View return policy
Description
BBS4 Monoclonal antibody specifically detects BBS4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BBS4 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI2D5 | |
| Western Blot | |
| Bardet-Biedl syndrome 4, Bardet-Biedl syndrome 4 protein | |
| Human recombinant protein fragment corresponding to amino acids 266-519 of human BBS4 (NP_149017) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Markers, Vision | |
| 585 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction