missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP2C1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-76566CL1
This item is not returnable.
View return policy
Description
ATP2C1 Polyclonal antibody specifically detects ATP2C1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| ATP2C1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| ATPase, Ca++ transporting, type 2C, member 1, BCPM, benign chronic pemphigus (Hailey-Hailey disease), Ca(2+)-sequestering, calcium-transporting ATPase type 2C member 1, EC 3.6.3, HHD, hSPCA1, HUSSY-28, PMR1L, secretory pathway Ca2+/Mn2+ ATPase 1 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ATP2C1. The immunogen is located within the last 50 amino acids of ATP2C1. Amino Acid Squence: IKKVERSREKIQKHVS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 27032 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction