missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Asprosin Antibody (Birdy-1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11677CL1
This item is not returnable.
View return policy
Description
Asprosin Monoclonal antibody specifically detects Asprosin in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Asprosin | |
| Monoclonal | |
| CoraFluor 1 | |
| FBN1, fibrillin 1 | |
| Recombinant human asprosin. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 2200 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Western Blot | |
| Birdy-1 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction