missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ANKZF1 Antibody [FITC], Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals™ NB100-61588F

Product Code. 30031674

  • £467.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ANKZF1 Polyclonal antibody specifically detects ANKZF1 in Human samples. It is validated for Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANKZF1
Polyclonal
PBS
Rabbit
Immunogen affinity purified
RUO
55139
Store at 4°C in the dark.
IgG
Immunoprecipitation, Western Blot
FITC
ankyrin repeat and zinc finger domain containing 1, ankyrin repeat and zinc finger domain-containing protein 1, FLJ13144, Zinc finger protein 744, ZNF744FLJ10415
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ankyrin repeat and zinc finger domain containing 1 using the numbering given in entry NP_060559.2 (GeneID 55139).
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato