missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMIGO Antibody (S86-36), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-22413CL1
This item is not returnable.
View return policy
Description
AMIGO Monoclonal antibody specifically detects AMIGO in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| AMIGO | |
| Monoclonal | |
| CoraFluor 1 | |
| adhesion molecule with Ig-like domain 1, ALI2, Alivin-2, AMIGO-1, AMIGOamphoterin-induced protein 1, amphoterin-induced gene and ORF, KIAA1163 | |
| Fusion protein amino acids 554-574 (cytoplasmic C-terminus) of human AMIGO-1 | |
| 0.1 mL | |
| Neuroscience | |
| 57463 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| S86-36 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction