missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1C1/AKR1C1 Antibody (AKR1C1/9069) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26923JF635
This item is not returnable.
View return policy
Description
Aldo-keto Reductase 1C1/AKR1C1 Monoclonal antibody specifically detects Aldo-keto Reductase 1C1/AKR1C1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Aldo-keto Reductase 1C1/AKR1C1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| 20 alpha-hydroxysteroid dehydrogenase, 20-ALPHA-HSD, 20-alpha-hydroxysteroid dehydrogenase, aldo-keto reductase family 1 member C1, aldo-keto reductase family 1, member C1 (dihydrodiol dehydrogenase 1; 20-alpha(3-alpha)-hydroxysteroid dehydrogenase), C9, Chlordecone reductase homolog HAKRC, DD1/DD2, DD1MGC8954, DDHH-37, dihydrodiol dehydrogenase 1, Dihydrodiol dehydrogenase 1/2, dihydrodiol dehydrogenase isoform DD1, EC 1.1.1, EC 1.1.1.-, EC 1.1.1.112, EC 1.1.1.149,2-ALPHA-HSD, EC 1.3.1.20, HAKRCDDH1aldo-keto reductase C, HBAB, hepatic dihydrodiol dehydrogenase, High-affinity hepatic bile acid-binding protein, Indanol dehydrogenase, MBAB, Trans-1,2-dihydrobenzene-1,2-diol dehydrogenase, type II 3-alpha-hydroxysteroid dehydrogenase | |
| Recombinant human full-length Aldo-keto Reductase 1C1/AKR1C1 protein | |
| 0.1 mL | |
| metabolism | |
| 1645 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin) | |
| AKR1C1/9069 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction