Learn More
TSC22D4 Antibody (3C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals H00081628-M06
Product Code. 18442379
  • £463.00 / 0.10mg
Description
TSC22D4 Monoclonal antibody specifically detects TSC22D4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSC22D4
Monoclonal
Unconjugated
In 1x PBS, pH 7.4
THG1, TILZ2THG-1, TSC22 domain family 4, TSC22 domain family protein 4, TSC22 domain family, member 4, Tsc-22-like protein THG-1, TSC22-related-inducible leucine zipper protein 2
TSC22D4 (AAH01966, 1 a.a. ∽ 395 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MSGGKKKSSFQITSVTTDYEGPGSPGASDPPTPQPPTGPPPRLPNGEPSPDPGGKGTPRNGSPPPGAPSSRFRVVKLPHGLGEPYRRGRWTCVDVYERDLEPHSFGGLLEGIRGASGGAGGRSLDSRLELASLGLGAPTPPSGLSQGPTSWLRPPPTSPGPQARSFTGGLGQLVVPSKAKAEKPPLSASSPQQRPPEPETGESAGTSRAATPLPSLRVEAEAGGSGARTPPLSRRKAVDMRLRMELGAPEEMGQVPPLDSRPSSPALYFTHDASLVHKSPDPFGAVAAQKFSLAHSMLAISGHLDSDDDSGSGSLVGIDNKIEQAMDLVKSHLMFAVREEVEVLKEQIRELAERNAALEQENGLLRALASPEQLAQLPSSGVPRLGPPAPNGPSV
0.1 mg
Cancer
81628
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
3C5
Western Blot 1:500, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 1:10-1:500
AAH01966
Mouse
IgG purified
RUO
Primary
Human
Purified