missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPK Antibody (2E10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00112858-M05
This item is not returnable.
View return policy
Description
PRPK Monoclonal antibody specifically detects PRPK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| PRPK | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| BUD32, C20orf64, chromosome 20 open reading frame 64, dJ101A2, dJ101A2.2, EC 2.7.11.1, Nori-2, Nori-2p, p53-related protein kinase, PRPK, TP53 regulating kinase, TP53-regulating kinase | |
| TP53RK (AAH09727, 154 a.a. ∽ 253 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. HDEDLIHGDLTTSNMLLKPPLEQLNIVLIDFGLSFISALPEDKGVDLYVLEKAFLSTHPNTETVFEAFLKSYSTSSKKARPVLKKLDEVRLRGRKRSMVG | |
| 0.1 mg | |
| Protein Kinase | |
| 112858 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 2E10 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence | |
| AAH09727 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction