missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEAK1 Antibody (1G9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00079834-M04
This item is not returnable.
View return policy
Description
PEAK1 Monoclonal antibody specifically detects PEAK1 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| PEAK1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| FLJ21140, FLJ34483, KIAA2002, NKF3 kinase family member, pseudopodium enriched atypical kinase, pseudopodium-enriched atypical kinase 1, SGK269, sugen kinase 269, tyrosine-protein kinase SgK269 | |
| PEAK1 (NP_079052.2, 1151 a.a. ∽ 1250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TPPPLPKKMIIRANTEPISKDLQKSMESSLCVMANPTYDIDPNWDASSAGSSISYELKGLDIESYDSLERPLRKERPVPSAANSISSLTTLSIKDRFSNS | |
| 0.1 mg | |
| Protein Kinase | |
| 79834 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 1G9 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| XP_370878 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction