missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MuRF1/TRIM63 Antibody (6G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084676-M01
This item is not returnable.
View return policy
Description
MuRF1/TRIM63 Monoclonal antibody specifically detects MuRF1/TRIM63 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA, KnockDown
Specifications
| MuRF1/TRIM63 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 6.3.2.-, IRFMURF2, Iris RING finger protein, MuRF1, MuRF-1, muscle specific ring finger protein 2, Muscle-specific RING finger protein 1, RING finger protein 28FLJ32380, RNF28E3 ubiquitin-protein ligase TRIM63, SMRZMURF-1, Striated muscle RING zinc finger protein, tripartite motif containing 63, tripartite motif-containing 63, Tripartite motif-containing protein 63 | |
| TRIM63 (NP_115977, 254 a.a. ∽ 352 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DKSTKLVETAIQSLDEPGGATFLLTAKQLIKSIVEASKGCQLGKTEQGFENMDFFTLDLEHIADALRAIDFGTDEEEEEFIEEEDQEEEESTEGKEEGH | |
| 0.1 mg | |
| Zinc Finger | |
| 84676 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA, KnockDown | |
| 6G6 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA, Knockdown Validated | |
| NP_115977 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction