missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGC20410 Antibody (1B11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00116071-M01
This item is not returnable.
View return policy
Description
MGC20410 Monoclonal antibody specifically detects MGC20410 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Specifications
| MGC20410 | |
| Monoclonal | |
| Unconjugated | |
| AAH12330 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 116071 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA | |
| 1B11 | |
| In 1x PBS, pH 7.4 | |
| basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
| BATF2 (AAH12330, 1 a.a. ∽ 189 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDCASCSAPGLLGCWDQAEGLLGPGPQGQHGCREQLELFQTPGSCYPAQPLSPGPQPHDSPSLLQCPLPSLSLGPAVVAEPPVQLSPSPLLFASHTGSSLQGSSSKLSALQPSLTAQTAPPQPLELEHPTRGKLGSSPDNPSSALGLARLQSREHKPALSAATWQGLVVDPSPHPLLAFPLLSSAQVHF | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction