missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG1 Kappa Light Chain, Clone: P3.6.2.8.1, PE, Isotype control, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43778-0.1mg
This item is not returnable.
View return policy
Description
IgG1 Kappa Monoclonal specifically detects IgG1 Kappa in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IgG1 Kappa Light Chain | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| Mouse | |
| Primary | |
| Mouse | |
| IgG1 κ |
| Flow Cytometry | |
| P3.6.2.8.1 | |
| PE | |
| Aqueous buffer, may contain carrier protein/stabilizer (proprietary). with 0.09% Sodium Azide | |
| 0.1 mg | |
| This monoclonal mouse IgG1, kappa immunoglobulin is useful as an isotype control. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction