missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK1 Antibody (4C2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00204851-M01
This item is not returnable.
View return policy
Description
HIPK1 Monoclonal antibody specifically detects HIPK1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Sandwich ELISA
Specifications
| HIPK1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| EC 2.7.11.1, homeodomain interacting protein kinase 1, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, KIAA0630MGC26642, MGC33446, MGC33548, Myak, Nbak2, nuclear body associated kinase 2b | |
| HIPK1 (AAH28408, 330 a.a. ∽ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. CTPLMVATLHPQVATITPQYAVPFTLSCAAGRPALVEQTAAVLQAWPGGTQQILLPSTWQQLPGVALHNSVQPTAMIPEAMGSGQQLADWRNAHSHGNQYS | |
| 0.1 mg | |
| Protein Kinase | |
| 204851 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 4C2 | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| AAH28408 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto