missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F4/80 Antibody (BM8), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-60140
This item is not returnable.
View return policy
Description
F4/80 Monoclonal specifically detects F4/80 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| F4/80 | |
| Monoclonal | |
| 0.1 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| ADGRE1 | |
| BALB/c macrophages obtained from 14-day-old bone marrow cell cultures | |
| 0.1 mg | |
| Immune System Diseases, Immunology, Innate Immunity, Myeloid derived Suppressor Cell | |
| 2015 | |
| Store at 4°C. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| BM8 | |
| Unconjugated | |
| ADGRE1, Adhesion G protein-coupled receptor E1, cell surface glycoprotein F4/80, DD7A5-7, EGF-like module containing, mucin-like, hormone receptor-like sequence 1, EGF-like module receptor 1, EGF-like module-containing mucin-like hormone receptor-like 1, EGF-TM7, EMR1 hormone receptor, F4/80, Gpf480, Ly71, lymphocyte antigen 71, Mouse EMR1, Rat EMR1, TM7LN3 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction