missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytochrome c Antibody (6H2.B4), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP2-24872
This item is not returnable.
View return policy
Description
Cytochrome c Monoclonal specifically detects Cytochrome c in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Block/Neutralize.
Specifications
| Cytochrome c | |
| Monoclonal | |
| 0.5 mg/mL | |
| Immunohistochemistry reported in scientific literature (PMID 28194435), Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16540654), Immunoprecipitation 2 ug/1X10^6 cells, Block/Neutralize | |
| CYCHCS, cytochrome c, cytochrome c, somatic, THC4 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody is reactive to Human, Mouse and Rat. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Blocking Assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 6H2.B4 | |
| Unconjugated | |
| P99999 | |
| CYCS | |
| Rat cytochrome c was used as immunogen to generate 6H2.B4 hybridoma clone (Goshorn et al, 1991). The antibody recognizes an epitope around residue 62 of rat Cytochrome c, based on competitive ELISA results (Goshorn et al. 1991). | |
| 0.1 mg | |
| Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers | |
| 54205 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction