missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N acetyl transferase 5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94531-0.02ml
This item is not returnable.
View return policy
Description
N acetyl transferase 5 Polyclonal antibody specifically detects N acetyl transferase 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| N acetyl transferase 5 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| dJ1002M8.1, dJ1175I6.2 (N-terminal acetyltransferase complex ard1 subunit), EC 2.3.1.88, homolog), N(alpha)-acetyltransferase 20, NatB catalytic subunit, N-acetyltransferase 3 homolog, N-acetyltransferase 5, N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae), N-acetyltransferase 5 (GCN5-related, putative), N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, N-alpha-acetyltransferase 20, NatB catalytic subunit, NAT3N-terminal acetyltransferase B complex catalytic subunit NAT5, NAT5dJ1002M8.1 (N-terminal acetyltransferase complex ard1 subunit), NatB complex subunit NAT5, N-terminal acetyltransferase B complex catalytic subunit NAA20, N-terminal acetyltransferase complex ARD1 subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 49-178 of human NAA20 (NP_057184.1). PGGELMGYIMGKAEGSVAREEWHGHVTALSVAPEFRRLGLAAKLMELLEEISERKGGFFVDLFVRVSNQVAVNMYKQLGYSVYRTVIEYYSASNGEPDEDAYDMRKALSRDTEKKSIIPLPHPVRPEDIE | |
| 0.02 mL | |
| Endocrinology | |
| 51126 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction