missing translation for 'onlineSavingsMsg'
Learn More

p70 S6 Kinase/S6K Antibody, Novus Biologicals™

Product Code. 18773274 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
Unit Size:
0.03mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18773274 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18773274 Supplier Novus Biologicals Supplier No. NBP223649SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 1 publication

p70 S6 Kinase/S6K Polyclonal specifically detects p70 S6 Kinase/S6K in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, Peptide ELISA, ICC/IF (Negative).
TRUSTED_SUSTAINABILITY

Specifications

Antigen p70 S6 Kinase/S6K
Applications Western Blot, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Peptide ELISA 1:100 - 1:1000, ICC/IF (Negative)
Formulation Whole sera with 50% Glycerol with 0.025% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, p70 ribosomal S6 kinase alpha, p70 S6 kinase alpha, p70 S6 kinase, alpha 1,70 kDa ribosomal protein S6 kinase 1, p70 S6 kinase, alpha 2, p70 S6KA, p70 S6K-alpha, p70(S6K)-alpha, p70-alpha, p70-S6K, P70S6K1, PS6K, ribosomal protein S6 kinase beta-1, Ribosomal protein S6 kinase I, ribosomal protein S6 kinase, 70kD, polypeptide 1, ribosomal protein S6 kinase, 70kDa, polypeptide 1, S6K, S6K1p70-S6K 1, S6K-beta-1, serine/threonine kinase 14 alpha, Serine/threonine-protein kinase 14A, STK14A
Gene Symbols RPS6KB1
Host Species Goat
Immunogen Synthetic peptide made to a C-terminal sequence of the human p70 S6 Kinase protein (between amino acids 350-472). [UniProt P23443-2].
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Autophagy, Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6198
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey and chimpanzee, 94% homologous in mouse and rat, 87% homologous in dog and 81% homologous in horse.
Target Species Human, Mouse, Rat, Bovine, Rabbit
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.