missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCL16 Antibody (4.4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50187-0.025ml
This item is not returnable.
View return policy
Description
CXCL16 Monoclonal antibody specifically detects CXCL16 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| CXCL16 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| chemokine (C-X-C motif) ligand 16, CXC chemokine ligand 16, CXCLG16, Scavenger receptor for phosphatidylserine and oxidized low density lipoprotein, SCYB16, Small-inducible cytokine B16, SRPSOXC-X-C motif chemokine 16, SR-PSOXTransmembrane chemokine CXCL16 | |
| GST-CXCL16 (143-224aa) fusion protein | |
| 0.025 mL | |
| Chemokines and Cytokines, Cytokine Research | |
| 58191 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| 4.4 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction