missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFYA Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93683-0.1ml
This item is not returnable.
View return policy
Description
NFYA Polyclonal antibody specifically detects NFYA in Human samples. It is validated for Western Blot
Specifications
| NFYA | |
| Polyclonal | |
| Western Blot 1:1000-1:2000 | |
| CAAT box DNA-binding protein subunit A, CBF-A, CBF-B, CCAAT-binding transcription factor subunit B, FLJ11236, HAP2, HAP2 CCAAT-binding protein, NF-YACAAT-box DNA binding protein subunit A, Nuclear transcription factor Y subunit A, nuclear transcription factor Y subunit alpha, nuclear transcription factor Y, alpha, Transcription factor NF-Y, A subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-318 of human NFYA (NP_068351.1). MEQYTANSNSSTEQIVVQAGQIQQQVQGQPLMVQVSGGQLITSTGQPIMVQAVPGGQGQTIMQVPVSGTQGLQQIQLVPPGQIQIQGGQAVQVQGQQGQTQQIIIQQPQTAVTAGQTQTQQQIAVQGQQVAQTAEGQTIVYQPVNADGTILQQVTVPVSGMITIPAASLAGAQIVQTGANTNTTSSGQGTVTVTLPVAGNVVNSGGMVMMVPGAGSVPAIQRIPLPGAEMLEEEPLYVNAKQYHRILKRRQARAKLEAEGKIPKERRKYLHESRHRHAMARKRGEGGRFFSPKEKDSPHMQDPNQADEEAMTQIIRVS | |
| 0.1 mL | |
| Cancer, Signal Transduction, Transcription Factors and Regulators | |
| 4800 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction