missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPK Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92676-0.1ml
This item is not returnable.
View return policy
Description
CENPK Polyclonal antibody specifically detects CENPK in Human samples. It is validated for Western Blot
Specifications
| CENPK | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| AF5alpha, CEMPK, CENP-KInterphase centromere complex protein 37, centromere protein K, FKSG14, ICEN37, leucine zipper protein FKSG14, P33, Protein AF-5alpha, SOLT, SoxLZ/Sox6-binding protein Solt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-269 of human CENPK (NP_071428.2). MNQEDLDPDSTTDVGDVTNTEEELIRECEEMWKDMEECQNKLSLIGTETLTDSNAQLSLLIMQVKCLTAELSQWQKKTPETIPLTEDVLITLGKEEFQKLRQDLEMVLSTKESKNEKLKEDLEREQRWLDEQQQIMESLNVLHSELKNKVETFSESRIFNELKTKMLNIKEYKEKLLSTLGEFLEDHFPLPDRSVKKKKKNIQESSVNLITLHEMLEILINRLFDVPHDPYVKISDSFWPPYVELLLRNGIALRHPEDPTRIRLEAFHQ | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 64105 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction